Learn More
Amperozide Dihydrochloride, TRC
Product Code. 30346182
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346182 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30346182 Supplier Toronto Research Chemicals Supplier No. TRCA63480325MG
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Amperozide Dihydrochloride
CAS 1307807-06-2
Molecular Formula C23H29F2N3O . 2(HCl)
Quantity 25 mg
Synonym 4-[4,4-bis(4-Fluorophenyl)butyl]-N-ethyl-1-piperazinecarboxamide Dihydrochloride, 4-[4,4-bis(4-Fluorophenyl)butyl]-N-ethyl-1-piperazinecarboxamide Hydrochloride (1:2)
InChI Formula InChI=1S/C23H29F2N3O.2ClH/c1-2-26-23(29)28-16-14-27(15-17-28)13-3-4-22(18-5-9-20(24)10-6-18)19-7-11-21(25)12-8-19;;/h5-12,22H,2-4,13-17H2,1H3,(H,26,29);2*1H
SMILES FC1=CC=C(C(C2=CC=C(F)C=C2)CCCN3CCN(C(NCC)=O)CC3)C=C1.Cl.Cl
IUPAC Name 4-(4,4-bis(4-fluorophenyl)butyl)-N-ethylpiperazine-1-carboxamide Dihydrochloride
Molecular Weight (g/mol) 474.41
Formula Weight 473.19