missing translation for 'onlineSavingsMsg'
Learn More

Amoxicillin sodium, MedChemExpress

Product Code. 16442189
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 16442189

Brand: Medchem Express HYB0467

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Amoxicillin (Amoxycillin) sodium is an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin sodium inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Amoxicillin (Amoxycillin) sodium is an antibiotic with good oral absorption and broad spectrum antimicrobial activity. Amoxicillin sodium inhibits the biosynthesis of polypeptides in the cell wall, thereby inhibiting cell growth.

Specifications

Chemical Name or Material Amoxicillin sodium
Physical Form Powder
CAS 34642-77-8
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C16H18N3NaO5S
For Use With (Application) COVID-19-immunoregulation
Synonym Amoxycillin sodium
Solubility Information H2O : 100 mg/mL (258.14 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (25.81 mM; Need ultrasonic)
SMILES O=C([C@@H]1N(C2=O)[C@]([C@@H]2NC([C@@H](C3=CC=C(O)C=C3)N)=O)([H])SC1(C)C)O[Na]
Molecular Weight (g/mol) 387.39
Formula Weight 387.39
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.