missing translation for 'onlineSavingsMsg'
Learn More

Ammonium copper(II) sulfate hexahydrate, 99%

Product Code. p-7020770
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11435233

Brand: Thermo Scientific Alfa Aesar A12834.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13587-26-3 | (NH4)2Cu(SO4)2·6H2O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13587-26-3
Molecular Formula (NH4)2Cu(SO4)2·6H2O
MDL Number MFCD00150440
Synonym Cupric ammonium sulfate

Specifications

Quantity 250 g
Formula Weight 399.83 (291.75 Anhydrous)
Percent Purity 99%
Chemical Name or Material Ammonium copper(II) sulfate hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.