missing translation for 'onlineSavingsMsg'
Learn More

Ammonium citrate dibasic, Honeywell™

Product Code. p-7145711 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
6 x 1 kg
Unit Size:
1kg
Pack of 6
This item is not returnable. View return policy

Product Code. 16615783

Brand: Honeywell Research Chemicals 25102H1KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., ≥98%

  • Base grade products, suitability tested for specific life science applications such as molecular biology or cell culture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3012-65-5
PubChem CID 13710713
ChEBI CHEBI:63076

Specifications

pH 4.5 - 5.5
Physical Form Solid
Quantity 1 kg
Percent Purity ≥98%
Grade Reagent
Packaging Plastic Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.