Learn More
Ammoninum-d4 Deuteroxide, TRC
Product Code. 30333062
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30333062 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30333062 Supplier Toronto Research Chemicals Supplier No. TRCA63402210G
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ammoninum-d4 Deuteroxide
CAS 12168-30-8
Molecular Formula D5NO
Quantity 10 g
Synonym Ammonium hydroxide ((ND4)(OD)) (8CI, 9CI), Ammoninum-d4 deuteroxide, Ammonium hydroxide D5, Ammonium hydroxide-d5
InChI Formula InChI=1S/H3N.H2O/h1H3;1H2/i/hD5
SMILES [2H]O[2H].[2H]N([2H])[2H]
Molecular Weight (g/mol) 40.08
Formula Weight 40.0685