Learn More
Amiselimod hydrochloride, MedChemExpress
Product Code. 16487156
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16487156
Brand: Medchem Express HY16734A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Amiselimod hydrochloride is a novel sphingosine 1-phosphate receptor-1 (S1P1) modulator, designed to reduce the bradycardia effects associated with fingolimod and other S1P receptor modulators.
Amiselimod hydrochloride is a novel sphingosine 1-phosphate receptor-1 (S1P1) modulator, designed to reduce the bradycardia effects associated with fingolimod and other S1P receptor modulators.
Specifications
Chemical Name or Material Amiselimod hydrochloride
CAS 942398-84-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H31ClF3NO3
Quantity 100 mg
Synonym MT-1303 hydrochloride
Solubility Information DMSO : 110 mg/mL (265.76 mM; Need ultrasonic)
SMILES OCC(CCC1=CC=C(OCCCCCCC)C(C(F)(F)F)=C1)(N)CO.[H]Cl
Molecular Weight (g/mol) 413.9
Formula Weight 413.9
Percent Purity 99.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death