Learn More
Aminoiminomethanesulfonic Acid, TRC
Product Code. 30327800
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30327800 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30327800 Supplier Toronto Research Chemicals Supplier No. TRCA6117555G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aminoiminomethanesulfonic Acid
CAS 1184-90-3
Molecular Formula C H4 N2 O3 S
Quantity 5 g
Synonym Formamidinesulfonic Acid, Aminoimino-methanesulfonic Acid, 1-Amino-1-imino-methanesulfonic Acid, AIMSA
InChI Formula InChI=1S/CH4N2O3S/c2-1(3)7(4,5)6/h(H3,2,3)(H,4,5,6)
SMILES NC(=N)S(=O)(=O)O
IUPAC Name amino(imino)methanesulfonic acid
Molecular Weight (g/mol) 124.12
Formula Weight 123.9943