missing translation for 'onlineSavingsMsg'
Learn More

AMG 487, MedChemExpress

Product Code. 16446736
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446736

Brand: Medchem Express HY15319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively.

AMG 487 is an orally active and selective antagonist of CXC chemokine receptor 3 (CXCR3) which inhibits the binding of CXCL10 and CXCL11 to CXCR3 with IC50s of 8.0 and 8.2 nM, respectively.

Specifications

Chemical Name or Material AMG 487
CAS 473719-41-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H28F3N5O4
Quantity 100 mg
Solubility Information DMSO : ≥ 41 mg/mL (67.93 mM)
SMILES O=C(N([C@@H](C1=NC2=NC=CC=C2C(N1C3=CC=C(OCC)C=C3)=O)C)CC4=CC=CN=C4)CC5=CC=C(OC(F)(F)F)C=C5
Molecular Weight (g/mol) 603.59
Formula Weight 603.59
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.