missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AMG 333

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AMG 333 is a potent and selective TRPM8 antagonist (IC50 values are 13 and 20 nM for hTRPM8 and rTRPM8, respectively). Displays high selectivity over other TRP channels (IC50 >20 μM, TRPV1/V3/V4/A1). Inhibits icilin -induced wet-dog shake in rats and suppresses cold-induced increases in blood pressure in the rat cold-pressor test.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C20H12F5N3O4 |
| Formula Weight | 453.33 |
| Product Type | Antagonist |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction