Learn More
Invitrogen™ AMFR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578768
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human Hela whole cell, human Daudi whole cell, rat kidney tissue. IHC: human placenta tissue, rat kidney tissue, mouse intestine tissue. ICC/IF: T47D cell. Flow: SiHa cell, U87 cell.
Autocrine motility factor is a tumor motility-stimulating protein secreted by tumor cells. The protein encoded by this gene is a glycosylated transmembrane protein and a receptor for autocrine motility factor. The receptor, which shows some sequence similarity to tumor protein p53, is localized to the leading and trailing edges of carcinoma cells.
Specifications
| AMFR | |
| Polyclonal | |
| Unconjugated | |
| AMFR | |
| AMF receptor; Amfr; Autocrine motility factor receptor; autocrine motility factor receptor, E3 ubiquitin protein ligase; E3 ubiquitin-protein ligase AMFR; gp78; RING finger protein 45; RING-type E3 ubiquitin transferase AMFR; RNF45 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23802, 267, 361367 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| Q9R049, Q9UKV5 | |
| AMFR | |
| E. coli-derived human AMFR recombinant protein (Position: E553-S643). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.