Learn More
Invitrogen™ Ambion™ RNA Century™ RNA Markers
Description
Ambion™ RNA Century™ Markers are unlabeled for staining with EtBr and are provided in one tube containing 50μg at a concentration of 1 mg/mL. Markers are premixed to give equivalent staining on a 5% polyacrylamide/8 M urea gel and are useful as RNA size standards in denaturing gel electrophoresis. 1–2μg (2–4μL) markers will generate 5 distinct bands on a TBE-Urea acrylamide gel with EtBr staining. Tested to be nuclease-free and stable overnight at 37°C. Compatible with denaturing agarose and denaturing polyacrylamide gels.
Acrylamide Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Concentration | 1 mg/mL |
| Form | Liquid |
| Quantity | 50 μg |
| Range | 100 to 500 bp |
| Product Type | RNA Marker |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.