Learn More
Invitrogen™ MagMAX™ for Stabilized Blood Tubes RNA Isolation Kit, compatible with Tempus™ Blood RNA Tubes
Product Code. 11330922
Click to view available options
Quantity:
96 preps
Unit Size:
Each
For Research Use Only. All usage must comply with product instructions.
Product Code. 11330922
Brand: Invitrogen™ 4451893
Please to purchase this item. Need a web account? Register with us today!
For Research Use Only. All usage must comply with product instructions.
Includes
Kit Components Box 1: Tempus™ Resuspension Solution, 12mL; Binding Solution Concentrate 5mL; Wash Solution 1 Concentrate, 2 x 16mL (Before using the Wash Solution 1 Concentrate, add the correct volume of isopropanol (user-supplied), as indicated on the bottle); Wash Solution 2 Concentrate, 30mL (Before using the Wash Solution 2 Concentrate, add the correct volume of ethanol (user-supplied), as indicated on the bottle); Elution Buffer 10mL; Processing Plate (96-well) 1 plate, Tempus™ Pre-Digestion Wash, 2 x 200mL; RNA Binding Beads, 2mL; Tempus™ 1X PBS, 2 x 150mL. Box 2: Tempus™ Proteinase, 0.25mL; TURBO™ DNase, 1mL
Requires
Refrigerated centrifuge (4°C) with swing bucket rotor capable of 5,000 x g; 50mL conical tube adapters; 50mL conical tubes; Tabletop microcentrifuge or picocentrifuge (for For tube format users); Blood tubes; Magnetic stand
The MagMAX for Stabilized Blood Tubes RNA Isolation Kit purifies total RNA (including miRNA) from Tempus Blood RNA Tubes using proven magnetic bead-based technology.
The MagMAX for Stabilized Blood Tubes RNA Isolation Kit purifies total RNA (including miRNA) from Tempus Blood RNA Tubes using proven magnetic bead-based technology. A TURBO DNase™ step efficiently eliminates contaminating gDNA ensuring the highest purity RNA possible suitable for real-time RT-PCR and microarray analysis.
Maximize Total RNA Yields From Blood Samples
The MagMAX for Stabilized Blood Tubes RNA Isolation Kit utilizes proven magnetic bead-based technology to purify maximal yields of total RNA from Tempus™ Blood RNA Tubes. The MagMAX for Stabilized Blood Tubes RNA Isolation Kit can recover 3-25 μg from a single Tempus Blood RNA tube. A TURBO DNase step efficiently eliminates contaminating gDNA that can interfere with downstream applications and ensures the highest purity RNA possible. The MagMAX for Stabilized Blood Tubes RNA Isolation Kit can also detect miRNA from Tempus Blood RNA tubes as shown by TaqMan™ gene expression assays. Users are strongly advised to perform appropriate validation studies against their miRNA targets.
Rapid Processing Time
Processing times will vary based on format selected, but twelve Tempus Blood RNA tubes can be processed manually with the MagMAX for Stabilized Blood Tubes RNA Isolation Kit in approximately 2 hours.
An Alternative to Full Purification
For users who perform primarily TaqMan-based real-time RT-PCR experiments, and do not need to archive RNA/cDNA, an alternative to the MagMAX for Stabilized Blood Tubes RNA Isolation Kit is the Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR. This kit makes it possible to perform expression analysis directly from Tempus Blood RNA tubes without conventional RNA purification.
One-Stop Shopping
MagMAX for Stabilized Blood Tubes RNA Isolation Kit is compatible with reverse transcription mixes (SuperScript™), PCR master mixes (both TaqMan and SYBRbased), and other blood workflow products.
  • Flexible purification format—use microfuge tubes, manual 96-well plates, or open-format automated 96-well plates from the same kit
  • Simple magnetic bead workflow—no spin columns
  • Total RNA without RNase activity, contaminating gDNA, or RT-PCR inhibitors
  • Suitable for use with automated protocols, high-throughput compatible
Flexible Purification Formats
  • The MagMAX for Stabilized Blood Tubes RNA Isolation Kit can be processed in low-, medium- or high-throughput formats with superior workflow and handling compared to traditional competitive methods
  • For low-throughput users, the MagMAX for Stabilized Blood Tubes RNA Isolation Kit can be used in individual 1.5 mL microfuge tubes with the bead-RNA-complex remaining in a single processing tube until the final elution step
  • For medium-to-high throughput users, the MagMAX for Stabilized Blood Tubes RNA Isolation Kit can be done in a 96-well processing plate with either a single or multi-channel pipettor to accelerate the purification process
  • For high-throughput users, the MagMAX for Stabilized Blood Tubes RNA Isolation Kit is compatible with manual and automated 96-well processing
  • Simply process multiple batches of tubes in series or in parallel, through the up-front centrifugation process, to the convenient stopping point, until enough samples can be loaded onto a 96-well processing plate for the RNA purification steps
  • The RNA purification process can be handled on either open-format liquid handlers, in conjunction with a 96-well magnetic stand on the deck, or magnetic bead movers like the MagMAX Express-96 Magnetic Particle Processor (deep well version only)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage
Box 1
• 12 mL Tempus Resuspension Solution; room temperature
• 5 mL Binding Solution (concentrated); room temperature
• 2 x 16 mL Wash Solution 1 (concentrated); room temperature
• 30 mL Wash Solution 2 (concentrated); room temperature
• 10 mL Elution Buffer; room temperature
• 1 Processing Plate; room temperature
• 2 x 200 mL Tempus Pre-Digestion Wash; 4°C
• 2 mL RNA Binding Beads; 4°C
• 2 x 150 mL Tempus 1X PBS; 4°C
Box 2
• 0.25 mL Tempus Proteinase; -20°C
• 1 mL TURBO DNase; -20°C
Isolation Technology Magnetic Bead
Purification Time Less than 2 hr. for automated protocol
Elution Volume 20 to 80 μL
Sample Type Whole Blood
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) KingFisher™ Duo Prime, KingFisher™ Flex
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity 96 preps
Shipping Condition Box 1: Room Temperature
Box 2: Cold Gel
Starting Material Amount 3 mL of whole blood collected in Tempus™ Blood RNA Tube
Yield ≤25 μg