missing translation for 'onlineSavingsMsg'
Learn More

AM-2394, MedChemExpress

Product Code. 16435597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435597 5 mg 5mg
16479506 50 mg 50mg
16434266 100 mg 100mg
16412858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16435597 Supplier Medchem Express Supplier No. HY100221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

Specifications

Chemical Name or Material AM-2394
CAS 1442684-77-6
Color White
Purity Grade Notes Research
Molecular Formula C22H25N5O4
Quantity 5 mg
Solubility Information DMSO : ≥ 30 mg/mL (70.84 mM)
SMILES O=C(NC)NC1=CC(OC2=CC(C)=CN=C2)=C(C3=CC=C(OCC(C)(O)C)N=C3)C=N1
Molecular Weight (g/mol) 423.47
Formula Weight 423.47
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.