missing translation for 'onlineSavingsMsg'
Learn More

AM-2394, MedChemExpress

Product Code. 16412858
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16412858

Brand: Medchem Express HY100221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

AM-2394 is a structurally distinct glucokinase activator (GKA). AM-2394 activates glucokinase (GK) with an EC50 of 60 nM.

Specifications

Chemical Name or Material AM-2394
CAS 1442684-77-6
Color White
Purity Grade Notes Research
Molecular Formula C22H25N5O4
Quantity 10 mg
Solubility Information DMSO : ≥ 30 mg/mL (70.84 mM)
SMILES O=C(NC)NC1=CC(OC2=CC(C)=CN=C2)=C(C3=CC=C(OCC(C)(O)C)N=C3)C=N1
Molecular Weight (g/mol) 423.47
Formula Weight 423.47
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.