Learn More
Lovibond™ Aluminum Vario Powder Set
Product Code. 10160823
Click to view available options
Quantity:
100 Pack
Unit Size:
Pack of 100
Product Code. 10160823
Brand: Lovibond™ undefined
Please to purchase this item. Need a web account? Register with us today!
Specifications
Quantity 100 Pack
compliance-icons compliance-icons
Product Identifier
  • vario aluminium set
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : ammonium fluoride, diammonium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate, methenamine