Learn More
Bel-Art™ SP Scienceware™ Teflon FEP Coated Aluminum Scoops
Product Code. 11731495 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
1 L
2 L
4 L
Unit Size:
Each
Product Code. 11731495
Brand: Bel-Art™ SP Scienceware™ H367330000
Only null left
Add to basket
Add to basket
Teflon FEP Coated Aluminum Scoops are FDA compliant for food contact
  • Anti-stick, corrosion resistant brown coating
  • Complies with FDA regulations for food contact
  • Handle is not coated so your grip is non-slip
Specifications
Description Sampling Scoop
Capacity (English) 1 qt.
Capacity (Metric) 1 L
Color Brown
Certifications/Compliance FDA Compliant for Food Contact
Product Type Scoop
Finish Teflon™
Width (Metric) 76 mm
Dimensions (L x W) 260 x 105 x 76 mm
Length (Metric) Handle 83 mm
Material Aluminum
Handle Type Non-Slip Grip
Length (Metric) 260 mm