missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, ≥99%, 100 to 200 Mesh, Honeywell Fluka™

Product Code. 15684130 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 15684130

Brand: Honeywell-Fluka 342653100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fused, powder, primarily α-phase, 100-200 mesh,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2040°C
Color Colorless
Quantity 100 g
Linear Formula Al2O3
UN Number NONH for all modes of transport
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96g/mol
Percent Purity ≥99%
Odor Odorless
Packaging Poly Bottle
Appearance Powder
Density 3.96g/cm3
Chemical Name or Material Aluminum oxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.