missing translation for 'onlineSavingsMsg'
Learn More

Korff™ Aluminum Foils

Brand:  Korff™ 60194N

Additional Details : Weight : 0.97200kg

 View more versions of this product

Product Code. 10140201

  • POA / 1 roll
Estimated Shipment: 03-07-2024
to see stock.

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »


Description

Description

Specifications

Specifications

10000 L x 300mm W
300mm
13 μm
Foil
Aluminum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions