Learn More
Bryson Packaging™ Aluminum Foil Rolls
Description
- Available in a variety of widths
- Can be cut to any length, cutter provided with box
- Sealing, weighing and transferring samples
Specifications
Specifications
| Applications | Sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes, suitable for use in autoclaves |
| Description | Aluminum foil |
| Item Description | Aluminum foil |
| Product Type | Aluminum Foil |
| Width (Metric) | 440mm |
| Material | Aluminum |
| Thickness | 10 μm |
| For Use With (Equipment) | Foil cutter |