Learn More
Bochem™ Aluminum Chemical Scoops
Product Code. 10769461
Click to view available options
Length (Metric):
135 mm
185 mm
210 mm
430 mm
Unit Size:
Each
Product Code. 10769461
Brand: Bochem™ 8818
Please to purchase this item. Need a web account? Register with us today!
Manufactured from aluminum. Bochem™ Aluminum Chemical Scoops comes in many sizes.
  • Chemical scoop
  • Manufactured from aluminum
  • Comes in many sizes
Specifications
Quantity 1 Ea.
Capacity (Metric) 90 mL
Product Type Chemical Scoop
Diameter (Metric) Outer 32mm
Material Aluminum
Length (Metric) 185 mm