missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Al/Si, AA 4643, A94643, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Aluminum, Al/Si, AA 4643, A94643, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Beryllium/Zirconium/Vanadium/Tin/Bismuth/Lead/Chromium/Gallium/Zinc/Copper/Nickel/Manganese/Titanium/Magnesium/Iron/Silicon/Boron/Aluminium/Calcium/Phosphorus |
| CAS | 7429-90-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Beryllium 0.004 %/Zirconium 0.008 %/Vanadium 0.01 %/Tin 0.017 %/Bismuth 0.019 %/Lead 0.02 %/Chromium 0.028 %/Gallium 0.028 %/Zinc 0.031 %/Copper 0.04 %/Nickel 0.04 %/Manganese 0.041 %/Titanium 0.052 %/Magnesium 0.1 %/Iron 0.16 %/Silicon 3.82 %/Boron (0.001) %/Aluminium (95.56) %/Calcium [0.005] %/Phosphorus [0.005] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction