missing translation for 'onlineSavingsMsg'
Learn More

Aluminum acetate, basic hydrate

Product Code. p-7081588
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11424713

Brand: Thermo Scientific Alfa Aesar A11620.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 142-03-0 | C4H7AlO5 | 162.08 g/mol

Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aluminum acetate, basic hydrate is used in medicine as an astringent and an antiseptic. It is also used as a mordant in dyeing, waterproofing and fireproofing agent. Further, it acts as a disinfectant in embalming and a dusting powder. In addition to this, it is used in antiperspirants.

Solubility
Insoluble in water

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-03-0
Molecular Formula C4H7AlO5
Molecular Weight (g/mol) 162.08
MDL Number MFCD00008688
InChI Key HQQUTGFAWJNQIP-UHFFFAOYSA-K
Synonym aluminum acetate, basic hydrate, c4h7alo5.h2o, aluminum diacetate hydrate
PubChem CID 18502856
SMILES CC(=O)O[Al](O)OC(C)=O

Specifications

Quantity 100 g
Merck Index 14,344
Solubility Information Insoluble in water
IUPAC Name diacetyloxyaluminum;hydrate
Chemical Name or Material Aluminum Acetate, Basic Hydrate, Basic hydrate
compliance-icons
Product Identifier
  • Aluminum acetate, basic hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aluminum acetate, basic hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.