missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Aluminum Reagent Powder Pillow Set

Product Code. 10430674
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10430674 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10430674 Supplier Hach™ Supplier No. 2603700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ready-to-use, premeasured powder reagents in individual sealed packets for determination of Aluminum by the Eriochrome Cyanine R (ECR) Method. Set contains ECR Reagent Powder Pillows, Hexamethylenetetramine Buffer Reagent Powder Pillows and ECR Masking Reagent Solution.

Powder Pillows make titration applications quick and easy.

  • Sealed packets ensure fresh, uncontaminated reagent every time
  • Premeasured to avoid measurement error and contamination from handling
  • Ready to use packets save time and hassle
  • Exact doses to easily monitor supply and prepare for lab procedures
  • Single-use packs for easy transportation and packing in test kits
TRUSTED_SUSTAINABILITY

Specifications

Analytes Aluminum
Format Individual packets and dropper bottle
For Use With (Application) Aluminum determination
For Use With (Equipment) DR6000, DR3900, DR1900
Includes ECR Reagent Powder Pillows, Hexamethylenetetramine Buffer Reagent Powder Pillows and ECR Masking Reagent Solution
No. of Tests 100
Quantity 1 Kit
Range 0.002 to 0.250mg/L Al
Sample Size 20 mL
Test Method Eriochrome Cyanine R (ECR)
Type Reagent kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.