Learn More
Alrestatin, MedChemExpress
Product Code. 16481098
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16481098
Brand: Medchem Express HYB1202
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Alrestatin is an inhibitor of aldose reductase, an enzyme involved in the pathogenesis of complications of diabetes mellitus, including diabetic neuropathy.
Alrestatin is an inhibitor of aldose reductase, an enzyme involved in the pathogenesis of complications of diabetes mellitus, including diabetic neuropathy.
Specifications
Chemical Name or Material Alrestatin
CAS 51411-04-2
Color White
Purity Grade Notes Research
Molecular Formula C14H9NO4
Quantity 5 mg
Synonym AY-22284
Solubility Information DMSO : 50 mg/mL (195.90 mM; Need ultrasonic)
SMILES O=C(O)CN(C(C1=CC=CC2=CC=CC3=C12)=O)C3=O
Molecular Weight (g/mol) 255.23
Formula Weight 255.23
Percent Purity 99.8%
Grade Research
Physical Form Solid