Learn More
Abnova™ ALPL/CEN1p FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | alkaline phosphatase, liver/bone/kidney |
| Gene Alias | AP-TNAP,FLJ40094,FLJ93059,HOPS,MGC161443,MGC167935,TNAP,TNSALP |
| Gene Symbol | ALPL |
| Gene ID (Entrez) | 249 |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |