Learn More
(AlphaZ)-Alpha-(Methoxyimino)-2-methylbenzeneacetic-d7 Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (alphaZ)-alpha-(Methoxyimino)-2-methylbenzeneacetic-d7 Acid Methyl Ester |
| CAS | 2734702-92-0 |
| Molecular Formula | C11H6D7NO3 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C11H13NO3/c1-8-6-4-5-7-9(8)10(12-15-3)11(13)14-2/h4-7H,1-3H3/b12-10-/i1D3,4D,5D,6D,7D |
| SMILES | [2H]C1=C(/C(C(OC)=O)=N/OC)C(C([2H])([2H])[2H])=C([2H])C([2H])=C1[2H] |
| IUPAC Name | methyl (Z)-2-(methoxyimino)-2-(2-(methyl-d3)phenyl-3,4,5,6-d4)acetate |
| Molecular Weight (g/mol) | 214.27 |
| Formula Weight | 214.13 |