Learn More
Ansell™ AlphaTec™ AquaDri™ Chemical Gloves
Product Code. 15355161 Shop All Ansell Products
Click to view available options
Size:
10
11
Unit Size:
12 pairs
Product Code. 15355161
Brand: Ansell™ 58330110
Please to purchase this item. Need a web account? Register with us today!
Two-layer construction keeps hands dryer for longer and with greater comfort and performance. Superior nitrile film provides advanced chemical protection against incidental exposure to bases, oils, fuels, some solvents, greases and animal fats.
  • Inner soft foam coating for moisture absorption
  • Nitrile film provides advanced chemical protection
  • Reversed-lozenge finish provides improved dry and wet grip
  • Nitrile compound for snag, puncture and abration protection
  • Foldable gutter cuff to prevent dripping on forearm
TRUSTED_SUSTAINABILITY
Specifications
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Palm Thickness (English) 0.0146 in.
Palm Thickness (Metric) 0.37 mm
Product Type Immersion Gloves
Size 11
Thickness (English) 0.0146 in.
Thickness (Metric) 0.3 mm
Glove Cuff Style Gauntlet
Liner Material Aquadri™ 2
Quantity 12 Pairs
Resistant To Chemical resistance
Series 58-330