Learn More
Ansell AlphaTec™ 2000 STANDARD Oversleeves Bound
Product Code. 16844800
Change view
Click to view available options
Size:
X-Large
Unit Size:
250 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16844800 X-Large 250 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16844800 Supplier Ansell Supplier No. WH20B0060005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Comfortable oversleeves featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Especificaciones
Size X-Large
Material Microporous Non-Woven Polyethylene Laminate
Color White
Seam Type Bound
Model WH20B
For Use With (Application) Cleanroom Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services
Certifications/Compliance (Europe) EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only
Weight (Metric) 63 gsm
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.