Learn More
(AlphaS,3S)-Alpha-[(tert-Butyloxycarbonyl)amino]-2-oxo-3-pyrrolidinepropanoic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | (alphaS,3S)-alpha-[(tert-Butyloxycarbonyl)amino]-2-oxo-3-pyrrolidinepropanoic Acid Methyl Ester |
| CAS | 328086-60-8 |
| Molecular Formula | C13H22N2O5 |
| Quantity | 50 mg |
| Synonym | (αS,3S)-α-[[(1,1-Dimethylethoxy)carbonyl]amino]-2-oxo-3-pyrrolidinepropanoic Acid Methyl Ester |
| InChI Formula | InChI=1S/C13H22N2O5/c1-13(2,3)20-12(18)15-9(11(17)19-4)7-8-5-6-14-10(8)16/h8-9H,5-7H2,1-4H3,(H,14,16)(H,15,18)/t8-,9-/m0/s1 |
| SMILES | O=C1[C@H](C[C@H](NC(OC(C)(C)C)=O)C(OC)=O)CCN1 |
| IUPAC Name | methyl (S)-2-((tert-butoxycarbonyl)amino)-3-((S)-2-oxopyrrolidin-3-yl)propanoate |
| Molecular Weight (g/mol) | 286.32 |
| Formula Weight | 286.15 |