missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin 3c Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81701H
This item is not returnable.
View return policy
Description
alpha Tubulin 3c Polyclonal antibody specifically detects alpha Tubulin 3c in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| alpha Tubulin 3c | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| Alpha-tubulin 2, Alpha-tubulin 3C/D, bA408E5.3, TUBA2Tubulin alpha-2 chain, TUBA3D, tubulin alpha-3C/D chain, tubulin, alpha 2, tubulin, alpha 3c | |
| Rabbit polyclonal alpha Tubulin 3c antibody was raised against a 16 amino acid peptide near the amino terminus of human alpha Tubulin 3c. The immunogen is located within amino acids 140 - 190 of alpha Tubulin 3c. Amino Acid Squence: MERLSVDYGKKSKLEF | |
| 0.1 mL | |
| Signal Transduction | |
| 7278 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction