Learn More
alpha-Propyl-alpha-(1-demethylethyl)-Verapamil Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride |
| CAS | 2732309-77-0 |
| Molecular Formula | C27 H38 N2 O4 . Cl H |
| Quantity | 10 mg |
| Synonym | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride, Verapamil Hydrochloride Imp. O (EP) as Hydrochloride, Benzeneacetonitrile','α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]methylamino]propyl]-3,4-dimethoxy-α-propyl-','hydrochloride (1:1) (ACI) |
| InChI Formula | InChI=1S/C27H38N2O4.ClH/c1-7-14-27(20-28,22-10-12-24(31-4)26(19-22)33-6)15-8-16-29(2)17-13-21-9-11-23(30-3)25(18-21)32-5;/h9-12,18-19H,7-8,13-17H2,1-6H3;1H |
| SMILES | Cl.CCCC(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2 |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propylpentanenitrile;hydrochloride |
| Molecular Weight (g/mol) | 491.06 |
| Formula Weight | 490.2598 |