Learn More
Alpha-Oxo-2-furanacetic Acid, TRC
Product Code. 30416210
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416210 1 g -
30424679 10 g -
30414374 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30416210 Supplier Toronto Research Chemicals Supplier No. TRCO8700301G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1467-70-5
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
Synonym 2-Furanacetic acid, α-oxo-, 2-Furanglyoxylic acid (7CI,8CI), α-Oxo-2-furanacetic acid, 2-(2-Furyl)-2-oxoacetic acid, 2-(Furan-2-yl)-2-oxoacetic acid, 2-Furanyl(oxo)acetic acid, 2-Furylglyoxylic acid, 2-Oxo-2-(2-furanyl)acetic acid, Furoylformic acid, NSC 59069
IUPAC Name 2-(furan-2-yl)-2-oxoacetic acid
SMILES OC(=O)C(=O)c1occc1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C6H4O4/c7-5(6(8)9)4-2-1-3-10-4/h1-3H,(H,8,9)
Formula Weight 140.011
Chemical Name or Material alpha-Oxo-2-furanacetic Acid