missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile, 98%, Thermo Scientific™

Product Code. 11425580
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425580 25 g 25g
11415580 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425580 Supplier Thermo Scientific Alfa Aesar Supplier No. H25844.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80866-87-1
Molecular Formula C10H8F3NO
Molecular Weight (g/mol) 215.175
MDL Number MFCD00001844
InChI Key UBOJBAYKXZRZHI-UHFFFAOYSA-N
Synonym 2-methoxy-2-phenyl-3,3,3-trifluoropropionitrile, alpha-methoxy-alpha-trifluoromethyl phenylacetonitrile, methoxyphenyl trifluoromethyl acetonitrile, acmc-1bl9n, -a-methoxy-a-trifluoromethylphenylacetonitrile, +/--a-methoxy-a-trifluoromethylphenylacetonitrile, #, +/--2-methoxy-2-trifluoromethyl phenylacetonitrile, +\--2-methoxy-2-phenyl-3,3,3-trifluoropropionitrile, alpha-trifluoromethyl-alpha-methoxybenzeneacetonitrile
PubChem CID 589652
IUPAC Name 3,3,3-trifluoro-2-methoxy-2-phenylpropanenitrile
SMILES COC(C#N)(C1=CC=CC=C1)C(F)(F)F

Specifications

Density 1.152
Boiling Point 76°C to 82°C (12 mmHg)
Flash Point 89°C (152°F)
Refractive Index 1.444
Quantity 25 g
UN Number UN3276
Formula Weight 215.18
Percent Purity 98%
Chemical Name or Material (+/-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.