missing translation for 'onlineSavingsMsg'
Learn More

alpha-Ethyl-3-nitrocinnamic acid, 99%, Thermo Scientific™

Product Code. p-7044785
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11378045

Brand: Thermo Scientific Alfa Aesar L08221.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124525-55-9
Molecular Formula C11H11NO4
Molecular Weight (g/mol) 221.212
MDL Number MFCD00134948
InChI Key QMUHBYNLXQHZNR-RMKNXTFCSA-N
Synonym e-2-3-nitrobenzylidene butanoic acid, alpha-ethyl-3-nitrocinnamic acid, 2e-2-3-nitrophenyl methylidene butanoic acid, 2-3-nitrobenzylidene butanoic acid, .alpha.-ethyl-3-nitrocinnamic acid, ^a-ethyl-3-nitrocinnamic acid, cinnamic acid, .alpha.-ethyl-m-nitro, butanoic acid, 2-3-nitrophenyl methylene, 2e-2-ethyl-3-3-nitrophenyl prop-2-enoic acid
PubChem CID 6059692
IUPAC Name (2E)-2-[(3-nitrophenyl)methylidene]butanoic acid
SMILES CCC(=CC1=CC(=CC=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 138°C to 142°C
Quantity 5 g
Beilstein 2618101
Formula Weight 221.21
Percent Purity 99%
Chemical Name or Material alpha-Ethyl-3-nitrocinnamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.