missing translation for 'onlineSavingsMsg'
Learn More

alpha-Boswellic acid, MedChemExpress

Product Code. 16451458
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16451458

Brand: Medchem Express HYN0611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress alpha-Boswellic acid (α-Boswellic acid) is a pentacyclic triterpene compound from extracts of Frankincense, has anticonvulsant and anti-cancer properties. alpha-Boswellic acid prevents and decreases the progression of Alzheimer’s hallmarks in vivo and can be used for Alzheimer’s disease research.

alpha-Boswellic acid (α-Boswellic acid) is a pentacyclic triterpene compound from extracts of Frankincense, has anticonvulsant and anti-cancer properties. alpha-Boswellic acid prevents and decreases the progression of Alzheimer’s hallmarks in vivo and can be used for Alzheimer’s disease research.

Specifications

Chemical Name or Material alpha-Boswellic acid
CAS 471-66-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H48O3
Quantity 5 mg
Synonym α-Boswellic acid
Solubility Information DMSO : 25 mg/mL (54.74 mM; Need ultrasonic)
SMILES C[C@@]([C@@]12C)(CC[C@@]3([H])[C@@]4(C)C(O)=O)[C@@](CC=C1[C@@](CC(C)(C)CC5)([H])[C@@]5(C)CC2)([H])[C@]3(CC[C@H]4O)C
Molecular Weight (g/mol) 456.7
Formula Weight 456.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.