missing translation for 'onlineSavingsMsg'
Learn More

Aloin B, TRC

Product Code. 30003685
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003685 25 mg 25mg
17912585 10 mg 10mg
17930975 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003685 Supplier Toronto Research Chemicals Supplier No. TRCA57541825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28371-16-6
Molecular Formula C21 H22 O9
Molecular Weight (g/mol) 418.39
Synonym 9(10H)-Anthracenone, 10-β-D-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, (10R)-, (10H)-Anthracenone, 10-β-D-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, (R)-, Anthrone, 10-glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-, stereoisomer (8CI), Isobarbaloin (7CI), (10R)-10-β-D-Glucopyranosyl-1,8-dihydroxy-3-(hydroxymethyl)-9(10H)-anthracenone, 10-epi-Aloin A, Aloin, Aloin B
IUPAC Name (10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-10H-anthracen-9-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14-,17-,19+,20-,21+/m1/s1
Formula Weight 418.1264
Chemical Name or Material Aloin B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.