missing translation for 'onlineSavingsMsg'
Learn More

Coolike Regeners™ Aloe Vera Body Cream

Product Code. 10224802
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10224802 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10224802 Supplier Coolike Regeners™ Supplier No. 7018 01000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Specifications

Product Type Aloe Vera Body Cream
Composition Panthenol and Q10
For Use With (Application) Sensitive and tired skin
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.