Learn More
Almorexant Hydrochloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Almorexant Hydrochloride |
| CAS | 913358-93-7 |
| Molecular Formula | C29H31F3N2O3. HCl |
| Quantity | 50 mg |
| Synonym | (alphaR,1S)-3,4-Dihydro-6,7-dimethoxy-N-methyl-alpha-phenyl-1-[2-[4-(trifluoromethyl)phenyl]ethyl]-2(1H)-Isoquinolineacetamide Monohydrochloride |
| InChI Formula | InChI=1S/C29H31F3N2O3/c1-33-28(35)27(20-7-5-4-6-8-20)34-16-15-21-17-25(36-2)26(37-3)18-23(21)24(34)14-11-19-9-12-22(13-10-19)29(30,31)32/h4-10,12-13,17-18,24,27H,11,14-16H2,1-3H3,(H,33,35)/t24-,27+/m0/s1 |
| SMILES | Cl.CNC(=O)[C@H](N1CCc2cc(OC)c(OC)cc2[C@@H]1CCc3ccc(cc3)C(F)(F)F)c4ccccc4 |
| IUPAC Name | (2R)-2-[(1S)-6,7-dimethoxy-1-[2-[4-(trifluoromethyl)phenyl]ethyl]-3,4-dihydro-1H-isoquinolin-2-yl]-N-methyl-2-phenylacetamide |
| Molecular Weight (g/mol) | 512.6 |
| Formula Weight | 548.2054 |