Learn More
Allyl Glycidyl Ether, TRC
Product Code. 30326291
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326291 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30326291 Supplier Toronto Research Chemicals Supplier No. TRCA55570010G
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[(2-Propen-1-yloxy)methyl]oxirane
CAS 106-92-3
Molecular Formula C6 H10 O2
Quantity 10 g
Synonym Oxirane, 2-[(2-propen-1-yloxy)methyl]-, Oxirane, [(2-propenyloxy)methyl]- (9CI), Propane, 1-(allyloxy)-2,3-epoxy- (6CI,8CI), 2-[(2-Propen-1-yloxy)methyl]oxirane, (±)-Allyl glycidyl ether, 1,2-Epoxy-3-allyloxypropane, 1-(2-Propenyloxy)-2,3-epoxypropane, 1-Allyloxy-2,3-epoxypropane, 2-(Allyloxymethyl)oxirane, 2-[(2-Propenyloxy)methyl]oxirane, 2-[(Prop-2-en-1-yloxy)methyl]oxirane, A 32608, AGE, AGE 500, Allyl 2,3-epoxypropyl ether, Allyl glycidyl ether, Allyl oxiranemethyl ether, Denacol EX 111, Glycidyl 2-propenyl ether, Glycidyl allyl ether, NSC 18596, NSC 631, Silong 191, [(2-Propenyloxy)methyl]oxirane, [(Allyloxy)methyl]oxirane
InChI Formula InChI=1S/C6H10O2/c1-2-3-7-4-6-5-8-6/h2,6H,1,3-5H2
SMILES C=CCOCC1CO1
IUPAC Name 2-(prop-2-enoxymethyl)oxirane
Molecular Weight (g/mol) 114.14
Formula Weight 114.0681