Learn More
Grace™ Alltech™ Fused Silica Cutter
Product Code. 10183474
Change view
Click to view available options
:
Tubing cutter; Ceramic fused silica
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10183474 Tubing cutter; Ceramic fused silica
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10183474 Supplier Grace™ Supplier No. 5122589
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Cleanly and squarely cuts fused-silica tubing for proper sealing with polyimide resin
Specifications
Type Tubing Cutter
For Use With (Equipment) Cleanly Cuts Ceramic Fused Silica Tubing of GC Capillary