missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Alloy Steel, Ni/Cr/Mo, AISI 8740, G87400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Lead/Titanium/Niobium/Arsenic/Vanadium/Tin/Cobalt/Nitrogen/Aluminium/Phosphorus/Copper/Molybdenum/Silicon/Carbon/Chromium/Nickel/Manganese/Zirconium/Antimony/Tungsten/Boron/Calcium/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.001 %/Titanium 0.001 %/Niobium 0.002 %/Arsenic 0.004 %/Vanadium 0.005 %/Tin 0.007 %/Cobalt 0.008 %/Nitrogen 0.0083 %/Aluminium 0.017 %/Phosphorus 0.025 %/Copper 0.109 %/Molybdenum 0.205 %/Silicon 0.248 %/Carbon 0.416 %/Chromium 0.501 %/Nickel 0.505 %/Manganese 0.92 %/Zirconium (<0.002) %/Antimony (<0.005) %/Tungsten (<0.005) %/Boron (0.0001) %/Calcium (0.0003) %/Oxygen (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction