missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, HY-TUF, A579 Grade 31, K32550, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, HY-TUF, A579 Grade 31, K32550, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Oxygen/Lead/Antimony/Titanium/Arsenic/Phosphorus/Cobalt/Nitrogen/Aluminium/Tin/Vanadium/Copper/Carbon/Chromium/Molybdenum/Manganese/Silicon/Nickel/Calcium/Magnesium/Zinc/Niobium/Zirconium/Tungsten |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0004 %/Oxygen 0.0006 %/Lead 0.0008 %/Antimony 0.0021 %/Titanium 0.0028 %/Arsenic 0.006 %/Phosphorus 0.006 %/Cobalt 0.008 %/Nitrogen 0.0102 %/Aluminium 0.019 %/Tin 0.021 %/Vanadium 0.023 %/Copper 0.11 %/Carbon 0.257 %/Chromium 0.38 %/Molybdenum 0.42 %/Manganese 1.37 %/Silicon 1.63 %/Nickel 1.76 %/Calcium (0.0001) %/Magnesium (0.0002) %/Zinc (0.001) %/Niobium (0.002) %/Zirconium (0.002) %/Tungsten (0.005) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction