missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Cr, AISI E52100, G52986, 50g, ARMI∣MBH
Click to view available options
Quantity:
50 g
Unit Size:
50g
Description
Alloy Steel, Cr, AISI E52100, G52986, 50g
Specifications
Specifications
| Chemical Name or Material | Arsenic/Nitrogen/Phosphorus/Tin/Molybdenum/Aluminium/Nickel/Vanadium/Copper/Silicon/Manganese/Carbon/Chromium |
| CAS | 7439-89-6 |
| Quantity | 50 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Arsenic 0.002 %/Nitrogen 0.002 %/Phosphorus 0.0049 %/Tin 0.0063 %/Molybdenum 0.0169 %/Aluminium 0.0258 %/Nickel 0.0411 %/Vanadium 0.0615 %/Copper 0.077 %/Silicon 0.246 %/Manganese 0.372 %/Carbon 1.023 %/Chromium 1.418 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction