Learn More
Allophycocyanin Antibody (936809) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8927P
Product Code. 18645624
  • 634.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Allophycocyanin Monoclonal antibody specifically detects Allophycocyanin in Broad Species Reactivity samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Allophycocyanin
Monoclonal
PE
PBS
Mouse
Protein A or G purified from cell culture supernatant
RUO
Broad Species Reactivity
Purified
Flow Cytometry, CyTOF
936809
Flow Cytometry, CyTOF-ready
APC
Allophycocyanin isolated from red algae, Accession # P25054
0.1 mL
Primary
Store at 4°C in the dark.
IgG2b