missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ All Purpose opener

Product Code. 10296562
Encompass
Change view
Click to view available options
Length (Metric):
260 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10296562 260 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10296562 Supplier Carl Roth™ Supplier No. 0506.1
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type All-Purpose Opener
Length (Metric) 260 mm
For Use With (Application) Used For Screw Lids from Tube Caps to Wide Neck Lids
No. per Pack 1 Ea.
Weight (Metric) 200g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.