Learn More
all-E-Heptaprenol, TRC
Specifications
Specifications
| Chemical Name or Material | all-E-Heptaprenol |
| CAS | 32304-16-8 |
| Molecular Formula | C35H58O |
| Quantity | 10 mg |
| Synonym | (all-E)-3,7,11,15,19,23,27-Heptamethyl-2,6,10,14,18,22,26-octacosaheptaen-1-ol |
| InChI Formula | InChI=1S/C35H58O/c1-29(2)15-9-16-30(3)17-10-18-31(4)19-11-20-32(5)21-12-22-33(6)23-13-24-34(7)25-14-26-35(8)27-28-36/h15,17,19,21,23,25,27,36H,9-14,16,18,20,22,24,26,28H2,1-8H3/b30-17+,31-19+,32-21+,33-23+,34-25+,35-27+ |
| SMILES | CC(=CCC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CC\C(=C\CO)\C)\C)\C)\C)\C)\C)C |
| IUPAC Name | (2E,6E,10E,14E,18E,22E)-3,7,11,15,19,23,27-heptamethyloctacosa-2,6,10,14,18,22,26-heptaen-1-ol |
| Molecular Weight (g/mol) | 494.83 |
| Formula Weight | 494.4488 |