missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALKBH7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15972745
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15972745 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15972745 Supplier Invitrogen™ Supplier No. PA576222

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ALKBH7 (alkB, alkylation repair homolog 7), also known as SPATA11, is a 221 amino acid protein belonging to the alkB family. ALKBH7 is one of many homologs of the Escherichia coli protein AlkB. AlkB functions to protect DNA and RNA against damage from environmental methylating compounds by directly reversing 1-methyladenine (1-meA) and 3-methylcytosine (3-meC) cytotoxic alkylation lesions in DNA and RNA. The enzyme acts by oxidative demethylation, utilizing ferrous iron and alpha-ketoglutarate as cofactors, 2-oxoglutarate as a co-substrate and molecular oxygen as the oxidizing agent. ALKBH7 is encoded by a gene located on human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognizedfor having the greatest gene density of the human chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH7
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene ALKBH7
Gene Accession No. Q9BT30, Q9D6Z0
Gene Alias 2310045B01Rik; 2510008E23Rik; Abh7; alkB homolog 7; alkB, alkylation repair homolog 7; alkB, alkylation repair homolog 7 (E. coli); Alkbh7; Alkylated DNA repair protein alkB homolog 7; alpha-ketoglutarate-dependent dioxygenase alkB homolog 7, mitochondrial; probable alpha-ketoglutarate-dependent dioxygenase ABH7; SPATA11; spermatogenesis associated 11; spermatogenesis cell proliferation related protein; spermatogenesis cell proliferation-related protein; Spermatogenesis-associated protein 11; UNQ6002; UNQ6002/PRO34564
Gene Symbols ALKBH7
Host Species Rabbit
Immunogen Recombinant fusion protein of human ALKBH7.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66400, 679944, 84266
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.