missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27973-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a homolog to the E. coli alkB gene product. The E. coli alkB protein is part of the adaptive response mechanism of DNA alkylation damage repair. It is involved in damage reversal by oxidative demethylation of 1-methyladenine and 3-methylcytosine. Dioxygenase that repairs alkylated single-stranded DNA and RNA containing 3-methylcytosine by oxidative demethylation. Requires molecular oxygen, alpha-ketoglutarate and iron. May have a role in placental trophoblast lineage differentiation. Has DNA lyase activity and introduces double-stranded breaks at abasic sites. Cleaves both single-stranded DNA and double-stranded DNA at abasic sites, with the greatest activity towards double-stranded DNA with two abasic sites.Specifications
| ALKBH1 | |
| Polyclonal | |
| Unconjugated | |
| ALKBH1 | |
| ABH, ABH1, alkB, ALKBH, ALKBH1, DNA lyase ABH1, hABH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8846 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13686 | |
| ALKBH1 | |
| ALKBH1 Fusion Protein Ag27520 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction