missing translation for 'onlineSavingsMsg'
Learn More

Hach™ Alkalinity Test Kit

Product Code. 10588924
Click to view available options
Range:
5 to 100mg/L or 20 to 400mg/L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10588924

Brand: Hach™ 2444301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Utilizes drop count titration method to measure alkalinity. Hach™ Alkalinity Test Kit, Model AL-AP, is a rugged kit that contains all required materials for 100 tests using drop count titration.

Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.

  • Utilizes drop count titration - a simple, economical titration method
  • Kit contains all required reagents and apparatus for 100 tests
  • All items included in a rugged carrying case for analysis in the field

Specifications

Analytes Alkalinity (as CaCO3)
Certifications/Compliance EPA
Description Model AL-AP
For Use With (Application) Alkalinity determination
Graduations 5mg/L or 50mg/L incremements
Includes Sample measuring tube, mixing bottle, reagents, instructions and carrying case
No. of Tests 100
Quantity 1 Kit
Range 5 to 100mg/L or 20 to 400mg/L
Test Method Drop count titration / Sulfuric acid
compliance-icons compliance-icons
Product Identifier
  • TEST KIT, AL-AP MG/L ALKALINITY
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H350i-May cause cancer by inhalation.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol, sulphuric acid, phenolphthalein
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.