missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Mouse anti-Human, Clone: 7G9E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60321-1-IG-150UL
This item is not returnable.
View return policy
Description
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3′ half of ALK, derived from chromosome 2, is fused to the 5′ portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.Specifications
| ALK/CD246 | |
| Monoclonal | |
| 1.6 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UM73 | |
| ALK | |
| ALK/CD246 Fusion Protein Ag21493 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| 7G9E3 | |
| Unconjugated | |
| ALK | |
| ALK, ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, CD246, TFG/ALK | |
| Mouse | |
| Protein A | |
| RUO | |
| 238 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction